Learn More
STA PHARMACEUTICAL US LLC 2'-OTBS azanebularine amidite | 200 mg | CAS 771494-03-2 | InChIKey QEYQVSAEEYPGQB-IEXCOMMKSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10157200MG
2'-OTBS azanebularine amidite is a Amidite reagent (Subcategory: Specialty Base Modification) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 771494-03-2
- MDL: No data
- InChIKey: QEYQVSAEEYPGQB-IEXCOMMKSA-N
- Molecular Weight: 870.075761998
- Molecular Formula: C45H60N7O7PSi
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-5-(3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C6=NC=NC=C6N=N5)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.