Learn More
STA PHARMACEUTICAL US LLC 2'-OTBS O6-NPE-G(di-Me) amidite | 1 g | CAS 2104647-87-0 | InChIKey ZEHMGNXQYCLVHJ-UTZPLLLRSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10184AK601GM
2'-OTBS O6-NPE-G(di-Me) amidite is a Amidite reagent (Subcategory: Guanine Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 2104647-87-0
- MDL: No data
- InChIKey: ZEHMGNXQYCLVHJ-UTZPLLLRSA-N
- Molecular Weight: 1077.304761998
- Molecular Formula: C56H73N8O10PSi
- Purity: ≥98%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-(dimethylamino)-6-(4-nitrophenethoxy)-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=C(N=C6OCCC7=CC=C([N+]([O-])=O)C=C7)N(C)C)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.