Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC 2'-OTBS O6-NPE-G(Me) amidite | 5 g | CAS 219648-40-5 | InChIKey DFBXNVVGLXNLNJ-UVQCDVNLSA-N

Supplier:  STA PHARMACEUTICAL US LLC STAAM10183AK1005GM

Encompass

2'-OTBS O6-NPE-G(Me) amidite is a Amidite reagent (Subcategory: Guanine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 219648-40-5
- MDL: No data
- InChIKey: DFBXNVVGLXNLNJ-UVQCDVNLSA-N
- Molecular Weight: 1063.277761998
- Molecular Formula: C55H71N8O10PSi
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-(methylamino)-6-(4-nitrophenethoxy)-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=C(N=C6OCCC7=CC=C([N+]([O-])=O)C=C7)NC)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C

Catalog No. 50-236-4223


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.