Learn More
STA PHARMACEUTICAL US LLC 2'-OTBS O6-NPE-G(Me) amidite | 200 mg | CAS 219648-40-5 | InChIKey DFBXNVVGLXNLNJ-UVQCDVNLSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10183200MG
2'-OTBS O6-NPE-G(Me) amidite is a Amidite reagent (Subcategory: Guanine Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 219648-40-5
- MDL: No data
- InChIKey: DFBXNVVGLXNLNJ-UVQCDVNLSA-N
- Molecular Weight: 1063.277761998
- Molecular Formula: C55H71N8O10PSi
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-(methylamino)-6-(4-nitrophenethoxy)-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=C(N=C6OCCC7=CC=C([N+]([O-])=O)C=C7)NC)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.