Learn More
Medchemexpress LLC 2-Oxazolidinone, 3-(1-oxo-2-buten-1-yl)-4-(phenylmethyl)-, (4S)- | 133812-16-5 | 99.5% | 245.27 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW013411
Encompass_Preferred
(E/Z)-Locostatin is a racemic mixture of Locostatin, which acts as a potent RKIP inhibitor. It functions by binding to the Raf kinase inhibitor RKIP protein, thereby disrupting the interaction between RKIP and Raf-1 kinase, as well as G protein-coupled receptor kinase 2. This compound has been shown to inhibit cell proliferation and migration and has been observed to worsen thioacetamide-induced acute liver failure in mice. It is intended for research use only and is not sold to patients.
  • Potent RKIP inhibitor.
  • Disrupts the interaction of RKIP with Raf-1 kinase and G protein-coupled receptor kinase 2.
  • Inhibits cell proliferation.
  • Inhibits cell migration.
Catalog No. 50-004-53201