Learn More
Medchemexpress LLC 2-oxazolidinone, 3-amino- | 80-65-9 | 98.0% | 102.09 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW012982100MG
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone, commonly detected as an indicator of furazolidone residues in vivo. This orally active compound is supplied as a solid, appearing colorless to off-white.
  • Metabolite of furazolidone
  • Indicator of furazolidone residues in vivo
  • Orally active
  • Appears as a solid, colorless to off-white
  • Inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver
  • Increases the levels of monoamine neurotransmitters in the brain
  • Lowers blood pressure in renal hypertensive rats
Catalog No. 50-003-67091