Learn More
Medchemexpress LLC 2-Oxazolidinone, 5-[(3,5-dimethylphenoxy)methyl]- | 1665-48-1 | 99.8% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0678
Encompass_Preferred
Metaxalone is an FDA-approved muscle relaxant that primarily acts on the central nervous system by inhibiting polysynaptic reflex arcs. It also functions as an inhibitor of MAO-A, possessing anti-inflammatory and antioxidant properties. It is for research use only and not sold to patients.
  • Inhibits polysynaptic reflex arcs
  • Functions as an inhibitor of MAO-A
  • Possesses anti-inflammatory properties
  • Possesses antioxidant properties
  • Can inhibit IL-1β-induced inflammatory phenotype
  • Modulates NF-κB and other related signaling pathways
  • Reduces MAO-A expression and activity in IL-1β-treated microglia
Catalog No. 50-004-06529