Learn More
Cayman Chemical 2-OxoadIpIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3457410 MG
Encompass_Preferred
An intermediate in the catabolism of L-tryptophan, L-lysine, and hydroxylysine; urinary excretion is increased in patients with α-ketoadipic aciduria
Catalog No. 50-294-7043