Learn More
Medchemexpress LLC 2-Oxosuccinic acid | 328-42-7 | MFCD00004245 | 99.9% | 132.07 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW010382R
Encompass_Preferred
Oxaloacetic acid (2-Oxosuccinic acid) is a metabolic intermediate involved in various pathways, including the citric acid cycle, gluconeogenesis, the urea cycle, the glyoxylate cycle, amino acid synthesis, and fatty acid synthesis. It facilitates the clearance of reactive oxygen species (ROS) and improves mitochondrial function.
  • Metabolic intermediate
  • Involved in citric acid cycle, gluconeogenesis, urea cycle, glyoxylate cycle, amino acid synthesis, and fatty acid synthesis
  • Facilitates clearance of reactive oxygen species (ROS)
  • Improves mitochondrial function
  • Ameliorates paraquat-induced acute lung injury by alleviating oxidative stress and mitochondrial dysfunction in mice (in vivo study)
  • Human endogenous metabolite
Catalog No. 50-004-53265