Learn More
Medchemexpress LLC 2-Oxovaleric acid | 1821-02-9 | 95.0% | 116.12 | 10 MM / 1 ML

Supplier: Medchemexpress LLC HY11309810MM/1ML
2-Oxovaleric acid is a keto acid found in human blood and is identified as an endogenous metabolite. This laboratory chemical is for research use only, intended for handling by qualified scientists in appropriately equipped and authorized facilities. It has not been fully validated for medical applications.
- Available in concentrations such as 10 mM in 1 mL DMSO.
- Purity of 95.0%.
- Molecular weight of 116.12.
- Provided as a colorless to light yellow liquid.
- Storage recommendations: 4°C protected from light; in solvent, -80°C for 6 months or -20°C for 1 month.
- Detailed documentation, including data sheet and SDS, is available.
- May cause skin irritation, serious eye irritation, and respiratory irritation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.