Learn More
Cayman Chemical 2-PalmItoyl Glycerol 25mg
SDP
Supplier:  Cayman Chemical 1788225 MG
Encompass_Preferred
A fatty acid ester that potentiates the activity of other endocannabinoids, including 2-AG to bind to CB1 and CB2 and inhibit adenylyl cyclase; interacts with endocannabinoids in the modulation of pain sensitivity
Catalog No. 50-296-6116