Learn More
Medchemexpress LLC 2-phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide | 113906-27-7 | 99.9% | 320.34 g·mol⁻¹ | C19H16N2O3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY101130200MG
Encompass_Preferred
PNU-74654 is a small-molecule inhibitor of the Wnt/β-catenin signaling pathway that disrupts the β-catenin-TCF interaction. It is supplied for research use and is characterized by reported cellular activity (IC50 129.8 μM in NCI-H295 cells), defined physicochemical properties, and high purity suitable for biochemical and cell-based assays.
  • Inhibits Wnt/β-catenin signaling by blocking β-catenin-TCF interaction.
  • Reported cellular potency: IC50 129.8 μM in NCI-H295 cells.
  • High chemical purity for reliable experimental results.
  • Well characterized physicochemical properties (MW 320.34, formula C19H16N2O3).
  • Suitable for in vitro biochemical and cell-based assays (research use only).
Catalog No. 50-002-13508