Learn More
Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g/mol | C9H12O | 10 G

Supplier: Medchemexpress LLC HYW01581010G
2-Phenyl-2-propanol (IUPAC: 2-phenylpropan-2-ol, CAS 617-94-7) is a tertiary alcohol used as a solvent and chemical intermediate. It is typically a liquid at room temperature, insoluble in water but soluble in common organic solvents, and is used in organic synthesis, fragrance ingredient production, and as a reaction regulator.
- Tertiary alcohol suitable as solvent and chemical intermediate.
- Molecular formula C9H12O; molecular weight 136.19 g/mol.
- Physical state liquid; insoluble in water, soluble in ethanol and ether.
- Boiling point about 202 °C; melting point about 36 °C.
- High purity grade (~99.5%) appropriate for research use.
- Common applications include fragrance additive synthesis and reaction regulation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.