Learn More
Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g·mol⁻¹ | C9H12O | 500 G

Supplier: Medchemexpress LLC HYW015810500G
2-Phenyl-2-propanol is an aromatic tertiary alcohol used as a chemical intermediate and biochemical reagent. It is commonly applied in organic synthesis (for example, in the preparation of diisopropylbenzene peroxide) and as an additive in fragrance and cosmetic formulations. The material is supplied in multiple pack sizes and is intended for research and formulation use.
- High purity: 99.5%.
- Molecular formula: C9H12O; molecular weight: 136.19 g·mol⁻¹.
- Available in solution and bulk pack sizes for flexibility in use.
- Storage stability: long-term frozen storage recommended for pure material.
- Suitable as a synthetic intermediate and formulation additive.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.