Learn More
Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD4456 | 99.5% | 136.19 g/mol | C9H12O | 100 G

Supplier: Medchemexpress LLC HYW015810100G
2-Phenyl-2-propanol is an aromatic tertiary alcohol used as a chemical intermediate and research reagent. It is employed in organic synthesis (for example, in the preparation of diisopropylbenzene peroxide) and as an additive in fragrance and cosmetic formulations. The material is supplied with specified purity and storage recommendations to preserve stability and performance.
- High purity (99.54%).
- Chemical formula C9H12O; molecular weight 136.19 g/mol.
- Appearance: colorless to off-white; solid below ~36°C, liquid above ~36°C.
- Available in common pack sizes including 100 g.
- Storage recommendations: pure form -20°C for long term, 4°C for medium term; in solvent -80°C for short term.
- Suitable as an intermediate in organic synthesis and for fragrance and cosmetic research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.