Learn More
Medchemexpress LLC 2-Phenylethanol | 60-12-8 | 99.99% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1290R50MG
Encompass_Preferred
2-Phenylethanol (Standard) is an analytical standard intended for research and analytical applications. It is an aromatic alcohol with a rose-like odor, functioning as a flavor and fragrance compound. It can be used as a preservative and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities. This analytical standard is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard
  • Aromatic alcohol with a rose-like odor
  • Functions as a flavor and fragrance compound
  • Can be used as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Utilized in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-11596