Learn More
Medchemexpress LLC 2-Phenylethanol (Standard) | 60-12-8 | 100.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1290R
Encompass_Preferred
2-Phenylethanol (Standard) is an analytical standard designed for research and analytical applications. This aromatic alcohol features a rose-like odor and functions as a flavor and fragrance compound. It also acts as a preservative and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for 2-Phenylethanol
  • Used in qualitative, quantitative, and methodological research experiments
  • Aromatic alcohol with a rose-like odor
  • Functions as a flavor and fragrance compound
  • Can be used as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
Catalog No. 50-004-14355