Learn More
Medchemexpress LLC 2-Phenylethyl β-D-thiogalactoside | 63407-54-5 | 99.9% | 300.37 | 100 MG

Supplier: Medchemexpress LLC HY137201100MG
2-Phenylethyl β-D-thiogalactoside is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It appears as a white to off-white solid with high purity.
- Used as a biochemical reagent
- Suitable for life science research
- High purity of 99.9%
- Soluble in DMSO (100 mg/mL)
- Stable at 4°C, stored under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.