Learn More
Medchemexpress LLC 2-Phenylethyl isothiocyanate-d5 | 912627-98-6 | 168.27 | C9H4D5NS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY23155S10MG
Encompass_Preferred
2-Phenylethyl isothiocyanate-d5 is the deuterated analogue of 2-phenylethyl isothiocyanate supplied as an analytical reference standard for research. It is intended for use as an internal standard in mass spectrometry and isotopic tracing studies, providing an isotopically labeled counterpart to the unlabeled analyte for accurate quantitation and method validation.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for isotopic tracing and quantitative analysis.
  • Chemical identity mirrors the unlabeled analyte for accurate quantitation.
  • Provided in small sealed vials appropriate for laboratory use.
  • Data sheet and handling instructions available.
Catalog No. 50-002-96862