Learn More
Medchemexpress LLC 2-Phenylglycine-d5 | 358731-96-1 | 99.0% | 156.19 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW010248S1MG
Encompass_Preferred
2-Phenylglycine-d5 is the deuterium labeled 2-Phenylglycine. 2-Phenylglycine (DL-α-Phenylglycine) is a metabolite in breast milk during the W2 to W4 lactation period. The product can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53752