Learn More
Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | MFCD00673896 | 99.2% | 308.37 g/mol | C19H20N2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10107410MM 1ML
Encompass_Preferred
UCM 608 (2-phenylmelatonin) is a high-affinity melatonin (MT) membrane receptor agonist used in receptor pharmacology and signaling studies. It is supplied as solid material or as a ready-to-use 10 mM solution in DMSO for use in binding and functional assays.
  • High affinity for MT1 and MT2 receptors (reported pKi values: MT1 10.7, MT2 10.4).
  • High purity suitable for research (99.18%).
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available in multiple solid quantities for dose flexibility.
  • Well characterized chemically (C19H20N2O2; molecular weight 308.37 g/mol; CAS 151889-03-1).
Catalog No. 50-002-13325