Learn More
Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | MFCD00673896 | 99.2% | 308.37 g/mol | C19H20N2O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101074100MG
Encompass_Preferred
UCM 608 (2-Phenylmelatonin) is a potent melatonin (MT) receptor agonist with high affinity for MT1 and MT2 receptors (reported pKi values ≈ 10.7 and 10.4). It is provided as a purified research-grade solid for use in in vitro pharmacology and receptor signaling studies.
  • High affinity for MT1 and MT2 receptors (pKi ≈ 10.7 and 10.4).
  • Potent agonist activity suitable for receptor pharmacology studies.
  • High purity (≈99.2%) for reliable experimental results.
  • Supplied as a solid in small research quantities for laboratory use.
  • Useful in studies of sleep, circadian biology, and receptor signaling.
Catalog No. 50-002-11193