Learn More
American Radiolabeled Chemicals Inc 2 PICOLINIC AC 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 182750UC
Encompass_Preferred
2-Picolinic acid-6-3H 50uCi in ethanol, 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 123.16, Exclusive
Catalog No. 50-901-244