Learn More
Bellco Glass, Inc. 2 Port Stainless Steel Assembly , 36L For Vertical Over Head Drive Vessels
SDP
Supplier:  Bellco Glass, Inc. 196423600R
Encompass_Preferred
2-port 45mm cap assembly, 36lFor 15l 1964 style Over Head Drive vessels. MADE IN THE USALong tube: 1/4"od x 18 1/4"Short tube: 1/4"od x 2.156"All tubes extended up 1.0" above the cap slugSupply 45mm s/cap & white silicone liner
Catalog No. 50-194-4781