Learn More
Medchemexpress LLC 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]- | 51384-51-1 | 99.9% | 267.36 | 10 MM / 1 ML
SDP
Supplier:  Medchemexpress LLC HY1750310MM/1ML
Encompass_Preferred
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Orally active
  • Functions as a selective β1-adrenoceptor antagonist
  • Shows anti-inflammatory effects
  • Exhibits antitumor properties
  • Demonstrates anti-angiogenic characteristics
Catalog No. 50-003-87184