Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol, 67-63-0, MFCD00011674, 4 L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 348634L
Encompass
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, for HPLC, 99.9%, Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol, isopropanol. 2-Propanol (Isopropanol) has been used for isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC. It has been used as solvent in the dissolution of Pt and TiO2 precursors, for the fabrication of Pt-TiO2/Ti electrodes.
Catalog No. NC0559120