Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propen-1-one, 3-(3,4-dihydroxy-2-methoxyphenyl)-1-(4-hydroxyphenyl)-, (2E)- | 58749-23-8 | MFCD20527294 | 99.9% | 286.28 | C16H14O5 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN037310MG

Encompass_Preferred

Licochalcone B is a naturally derived chalcone used as a research reagent to investigate amyloid-β aggregation, inflammasome signaling, and apoptosis pathways. It is provided as a light yellow solid with high analytical purity for biochemical and cell-based assays.

  • High analytical purity (99.91% by LCMS)
  • Molecular formula C16H14O5; molecular weight 286.28
  • CAS number 58749-23-8
  • Light yellow to yellow solid appearance
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month)
  • Available in solid and DMSO solution formats (10 mM, 1 mL)
  • Intended for biochemical and cell-based research applications

Catalog No. 50-002-96272


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.