Learn More
Medchemexpress LLC 2-Propen-1-one, 3-(3-pyridinyl)-1-(4-pyridinyl)- | 13309-08-5 | 99.9% | 210.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1101691MG
Encompass_Preferred
(E/Z)-3PO is a potent PFKFB3 inhibitor available as an off-white to light yellow solid. This compound has a molecular formula of C13H10N2O and a molecular weight of 210.24, with a high purity of 99.9% by LCMS. It is intended for laboratory research use only.
  • Potent PFKFB3 inhibitor
  • High purity for reliable research
  • Off-white to light yellow solid form
  • Suitable for various laboratory applications
Catalog No. 50-003-44786