Learn More
Medchemexpress LLC 2-Propenoic acid, 3-(4-hydroxy-3-methoxyphenyl)-, (2S)-2-hydroxy-2-(4-hydroxyphenyl)ethyl ester | 272122-56-2 | 1 MG

Supplier: Medchemexpress LLC HYN37011MG
Decursidate is a chemical compound supplied as a white to off-white solid powder. It has a molecular formula of C18H18O6 and a molecular weight of 330.33. The purity is determined by LCMS to be 98.08%.
- White to off-white solid
- Molecular formula: C18H18O6
- Molecular weight: 330.33
- Purity (LCMS): 98.08%
- Storage conditions for powder: -20°C for 3 years
- Storage conditions in solvent: -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.