Learn More
Medchemexpress LLC 2-propenoic acid, homopolymer | 9003-01-4 | MFCD00084394 | 450,000 (average) | (C3H4O2)X | 10 G
SDP
Supplier:  Medchemexpress LLC HYW243303E10G
Encompass_Preferred
Poly(acrylic acid) (average Mw 450,000) is a high-molecular-weight anionic polymer used in research and formulation applications where surface stabilization, thickening, or corrosion mitigation is required. It is supplied as a white to off-white solid and should be stored sealed, away from moisture.
  • High average molecular weight for effective viscosity and film formation.
  • Anionic polyelectrolyte useful for stabilizing colloids and suspensions.
  • Compatible with aqueous systems and common laboratory solvents.
  • Suitable for corrosion inhibition, coatings, and drug delivery research.
  • Solid form simplifies handling and preparation of small laboratory quantities.
Catalog No. 50-002-82500