Learn More
Medchemexpress LLC 2-pyrazinecarboxamide, 6-ethyl-5-[(trans-4-methoxycyclohexyl)amino] | 1373409-08-5 | 98.0% | 580.76 g/mol | C31H48N8O3 | 5 MG

Supplier: Medchemexpress LLC HY1117525MG
EML4-ALK kinase inhibitor 1 is a potent, orally active small-molecule inhibitor of the EML4-ALK fusion kinase. Reported biochemical potency is IC50 = 1 nM, and the compound shows cellular antiproliferative activity in BaF3 cells expressing EML4-ALK. Supplied as a high-purity research reagent for in vitro and in vivo studies.
- Potent biochemical inhibition (IC50 1 nM).
- Demonstrated antiproliferative activity in EML4-ALK BaF3 cells (~9.7 nM).
- High purity 98.0%.
- Molecular weight 580.76 g/mol; formula C31H48N8O3.
- Soluble in DMSO (110 mg/mL) and compatible with reported in vivo formulations.
- Available in milligram pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.