Learn More
Medchemexpress LLC 2-Pyrazinecarboxamide, 6-ethyl-5-[(trans-4-methoxycyclohexyl)amino]-3-[[3-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]amino]- | 1373409-08-5 | 98.0% | 580.76 | 50 MG
Supplier: Medchemexpress LLC HY11175250MG
EML4-ALK kinase inhibitor 1 is a potent, orally active inhibitor of echinoderm microtubule-associated protein-like 4-anaplastic lymphoma kinase (EML4-ALK), with an IC50 of 1 nM. It is for research use only and not sold to patients.
- Potent orally active inhibitor of EML4-ALK with an IC50 of 1 nM.
- Available in various sizes, including free samples.
- Provided with detailed solubility and storage information for both in vitro and in vivo applications.