Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid | 98-97-5 | 99.9% | C5H4N2O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HY76353100G
Encompass_Preferred
2-Pyrazinecarboxylic acid is a biochemical reagent suitable for life science-related research, serving as a biological material or organic compound. It is known as an active metabolite of pyrazinamide, an anti-tubercular agent. Studies show its urinary excretion can be decreased by probenecid, and in vitro, it increases urate uptake in rat renal brush-border membrane vesicles.
  • Active metabolite of pyrazinamide, an anti-tubercular agent
  • Can be used as a biological material or organic compound for life science research
  • Suppresses 1-aminocyclopropane-1-carboxylic acid oxidase (ACO) activity in plant cells
Catalog No. 50-004-34137