Learn More
Medchemexpress LLC 2-pyridinecarboxamide | 1452-77-3 | MFCD00023483 | 100.0% | 122.13 | C6H6N2O | 10 G
SDP
Supplier:  Medchemexpress LLC HY10102010G
Encompass_Preferred
Picolinamide (2-pyridinecarboxamide) is a small-molecule research reagent reported to inhibit poly(ADP-ribose) synthetase (PARS) and is supplied as a solid for laboratory use. The compound is identified by CAS number 1452-77-3 and is intended for research applications only.
  • Small-molecule pyridinecarboxamide for research use.
  • Reported inhibitor of poly(ADP-ribose) synthetase (PARS).
  • Available as a solid in multiple pack sizes, including 10 G.
  • High purity suitable for analytical and biological assays.
  • Molecular weight 122.13 g/mol; formula C6H6N2O.
Catalog No. 50-002-11309