Learn More
Medchemexpress LLC 2-Pyridinecarboxamide, 5-[2-(5-isoquinolinyl)ethynyl]-N-[4-(4-morpholinylmethyl)-3-(trifluoro... | 2803422-60-6 | 98.2% | 516.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY171230100MG
Encompass_Preferred
MTH1 activator-1 is an MTH1 activator that enhances endogenous MTH1 activity and significantly reduces 8-oxo-dG levels in cellular DNA. It can be used to probe cellular and biological effects of upregulated oxidative damage repair in nucleotide pools and to delay or abrogate tumorigenesis. This product is for research use only.
  • Enhances endogenous MTH1 activity
  • Significantly reduces 8-oxo-dG levels in cellular DNA
  • Used to probe cellular and biological effects of upregulated oxidative damage repair
  • May delay or abrogate tumorigenesis
  • Available as a white to off-white solid
  • Recommended storage conditions for powder include -20°C for 3 years or 4°C for 2 years
  • Recommended storage conditions in solvent include -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-86354