Learn More
Medchemexpress LLC 2-pyridinecarboxamide, N-[4-[[3-[(1-oxo-2-propen-1-yl)amino]benzoyl]amino]phenyl]-6-(1H-pyrazol-3-yl) | 2227368-54-7 | MFCD34593626 | 99.0% | 452.46 g·mol-1 | C25H20N6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY103017A10MG
Encompass_Preferred
JH-X-119-01 is a small-molecule inhibitor selective for interleukin-1 receptor-associated kinase 1 (IRAK1). It demonstrates potent biochemical inhibition (IC50 ≈ 9 nM), shows no measurable IRAK4 inhibition up to 10 μM, and has reported efficacy in LPS-induced sepsis models. The compound is supplied as a high-purity research reagent in solid form or as a DMSO solution for experimental use.
  • Potent IRAK1 inhibition (IC50 ≈ 9 nM).
  • No measurable IRAK4 inhibition up to 10 μM.
  • Demonstrated in vivo efficacy in LPS-induced sepsis models.
  • High supplied purity (≈99.0%).
  • Available as solid and 10 mM DMSO solution for easy dosing.
  • Intended for research use only.
Catalog No. 50-002-47607