Learn More
Medchemexpress LLC 2-Pyridinecarboxylic acid, 4-bromo | 30766-03-1 | 202.007 | 10 G

Supplier: Medchemexpress LLC HYW00486010G
4-Bromopicolinic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a useful synthetic intermediate and is used as an intermediate for agrochemical, pharmaceutical, and dyestuff fields.
- Used as a biological material or organic compound for life science research.
- Useful synthetic intermediate for agrochemical, pharmaceutical, and dyestuff fields.
- Appearance: White to light yellow solid.
- Purity: 99.89% (HPLC).
- Storage conditions (powder): -20°C for 3 years, 4°C for 2 years.
- Storage conditions (in solvent): -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.