Learn More
Medchemexpress LLC 2-(pyridyldithio)ethylamine hydrochloride | 106139-15-5 | MFCD01863453 | 99.9% | 222.76 g/mol | C7H11ClN2S2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY101794200MG
Encompass_Preferred
2-(Pyridyldithio)ethylamine hydrochloride is a disulfide intercalating cross-linking reagent that combines a 2-pyridyl disulfide moiety with a primary amine, supplied as a hydrochloride salt. It is designed for introducing cleavable disulfide handles in bioconjugation and reversible cross-linking applications, supporting peptide and protein conjugation workflows with high purity and defined storage recommendations.
  • Enables formation of reversible disulfide linkages for cleavable conjugates.
  • Provides a reactive primary amine for standard coupling chemistries.
  • Contains a 2-pyridyl disulfide for efficient thiol-exchange reactions.
  • Suitable for peptide and protein conjugation workflows.
  • High purity supports reproducible conjugation results.
Catalog No. 50-002-48770