Learn More
Medchemexpress LLC 2-Pyrimidinamine, 5-[8-methyl-9-(1-methylethyl)-2-(4-morpholinyl)-9H-purin-6-yl]- | 1246560-33-7 | 98.7% | 354.41 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16585100MG
Encompass_Preferred
VS-5584 is a pan-PI3K/mTOR kinase inhibitor that simultaneously blocks both mTORC2 and mTORC1. This ATP-competitive inhibitor selectively targets PI3K/mTOR signaling with low nanomolar potency against all human Class I PI3K isoforms and mTOR kinase. It preferentially inhibits cancer stem cells in HMLE breast cancer cells, reducing Aldefluor-positive cancer stem cells. In vivo, it reduces cancer stem cells in breast cancer tumors and induces tumor regression in Docetaxel-resistant models. Rapidly absorbed with a tmax of 0.9 hours and an elimination half-life of 10 hours, it leads to significant tumor growth inhibition.
  • Pan-PI3K/mTOR kinase inhibitor
  • Blocks mTORC2 and mTORC1
  • Selectively inhibits PI3K/mTOR signaling
  • Inhibits cancer stem cells
  • Induces tumor regression
  • Rapidly absorbed, long half-life
  • Leads to tumor growth inhibition
Catalog No. 50-003-93902