Learn More
Medchemexpress LLC 2-(Pyrrolidin-1-yl)ethanol | 2955-88-6 | 99.6% | 115.18 | 25 G

Supplier: Medchemexpress LLC 25GHYW010519
2-(Pyrrolidin-1-yl)ethanol is a biochemical reagent suitable for use as a biological material or organic compound in various life science related research applications.
- Can be used as a biological material
- Organic compound for life science research
- Appearance: liquid
- Color: colorless to light yellow
- Molecular formula: C6H13NO
- Molecular weight: 115.18
- Soluble in DMSO (200 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.