Learn More
Medchemexpress LLC 2-pyrrolidinemethanol, 1-methyl-, (2S)- | 34381-71-0 | MFCD00011727 | 99.3% | 115.17 g/mol | C6H13NO | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW019679
Encompass_Preferred
(S)-N-Methylpyrrolidine-2-methanol is a chiral small-molecule research reagent used as the (S)-enantiomer of N-methyl-2-pyrrolidinemethanol. It is employed as a building block, intermediate, or reference standard in organic synthesis and life-science research, and is provided with analytical documentation validating purity and identity.
  • Chiral (S)-enantiomer suitable for stereospecific synthesis.
  • High purity (99.3% by GC) for analytical and synthetic applications.
  • Chemical formula C6H13NO; molecular weight 115.17 g/mol.
  • Commonly used as a building block, intermediate, or reference standard.
  • Available in 25 g and 50 g packages for laboratory-scale work.
Catalog No. 50-003-03001