Learn More
Medchemexpress LLC 2-((R)-1-((R)-2-(3-((1,1-difluoro-2-(pyridin-2-yl)ethoxy)methyl)phenyl)-2-hydroxyethyl)-2,3-dihydro-1H-inden-2-yl)acetic acid | 870964-67-3 | 99.28% | 374.48 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1761310MM/1ML
Encompass_Preferred
Evocalcet (KHK7580) is an orally active agonist of the calcium sensing receptor (CaSR) that inhibits the secretion of parathyroid hormone (PTH) from parathyroid gland cells. It can be utilized in the research of hyperparathyroidism.
  • Orally active CaSR agonist.
  • Inhibits parathyroid hormone secretion.
  • Potentially useful for hyperparathyroidism research.
  • Dose-dependently increases intracellular calcium with an EC50 value of 92.7 nM in hCaR-HEK293 cells.
  • Shows no significant inhibitory effects on specific activities of cytochrome P450 isozymes.
  • Reduces serum parathyroid hormone and calcium levels in animal models.
  • Increases serum inorganic phosphorus level in animal models.
Catalog No. 50-003-87306