Learn More
Medchemexpress LLC 2-sec-butyl-3-methoxypyrazine | 24168-70-5 | MFCD00006136 | 99.0% | 166.22 g/mol | C9H14N2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01714010MM/1ML
Encompass_Preferred
2-Sec-butyl-3-methoxypyrazine (SBMP) is a methoxypyrazine research reagent provided as a 10 mM solution in DMSO. It is commonly used as an analytical standard and in studies of flavor, fragrance, and olfactory-active compounds; it occurs naturally in certain insects and plants. For research use only.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity supporting analytical applications.
  • Suitable as analytical standard for flavor and fragrance research.
  • Useful in olfactory, sensory, and biological studies.
  • Supplied in 1 mL volume for small-scale experiments.
Catalog No. 50-003-01161