Learn More
Medchemexpress LLC 2-Selenouracil | 16724-03-1 | 98.0% | 175.05 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11195910MG
Encompass_Preferred
2-Selenouracil is a specialized photosensitizer used for photodynamical therapy.
  • Specialized photosensitizer for photodynamical therapy.
  • Targets reactive oxygen species (ROS).
  • Involved in immunology/inflammation, metabolic enzyme/protease, and NF-κB pathways.
  • Powder storage at -20°C for 3 years or 4°C for 2 years.
  • In solvent storage at -80°C for 6 months or -20°C for 1 month.
  • Soluble in ethanol (2.5 mg/mL with ultrasonic) and slightly soluble in DMSO (< 1 mg/mL with ultrasonic, warming, and heating to 60°C).
  • For research use only.
Catalog No. 50-004-36830