Learn More
Elemental Scientific Inc 2 STP PVC TBG 1.02MM 18IN PK12
SDP
Supplier:  Elemental Scientific Inc ES43971102
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 2-stop, 1.02mm ID, 2.0mm flared 151mm bridge distance (white/white), 12 pc.
Catalog No. NC0567609