Learn More
AVANTOR MASTERFLEX (VWR LLC) 2-STP TYGN E-LAB 2.29MMID 12PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9646044
Encompass
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 2.29 mm ID; 12/PK, Avantor
Catalog No. 50-274-1917