Learn More
Apexbio Technology LLC 2-Thio-dTTP(Synonyms: 2-Thio-2'-deoxythymidine-5'-triphosphate, 2-Thio-dTTP, 2-Thio-thymidine-5'-triphosphate), 10ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B81000.01
Encompass_Preferred
2-Thio-dTTP (2-thio-2 -deoxythymidine-5 -triphosphate) is a modified nucleotide analog in which the 2-position oxygen atom of thymine is substituted by sulfur Incorporation of this modified nucleotide during DNA synthesis can influence DNA-protein interactions and alter nucleic acid stability It is commonly utilized in molecular biology studies involving DNA replication fidelity mutagenesis and structural analyses of nucleic acids Due to the sulfur substitution 2-Thio-dTTP finds applications in biochemical assays and DNA sequencing methodologies
Catalog No. 50-000-60735