Learn More
Medchemexpress LLC 2-Thiooxamic acid ethyl ester | 16982-21-1 | MFCD00040713 | 99.5% | C4H7NO2S | 100 G

Supplier: Medchemexpress LLC HY78216100G
2-Thiooxamic acid ethyl ester is an organic compound that contains both thiocarbonyl and ethyl ester functional groups. This compound has applications in scientific research, specifically in the study of various types of cancer, including chronic myeloid leukemia and breast cancer.
- An organic compound.
- Contains thiocarbonyl and ethyl ester groups.
- Purity of 98.84%.
- Appears as an off-white to yellow solid.
- Soluble in DMSO.
- Used in the study of cancer.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.