Learn More
Medchemexpress LLC 2-Thiophenemethanol | 636-72-6 | 99.97% | 114.17 | 50 G

Supplier: Medchemexpress LLC HY3475150G
2-Thiophenemethanol is a biochemical reagent suitable for life science research. It is intended for research use only.
- Can be used as a biological material or organic compound
- Ships at room temperature in continental US
- Store at 4°C, protect from light
- Soluble in DMSO (200 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.