Learn More
Medchemexpress LLC 2-thioxo-5-((5-(3-(trifluoromethyl)phenyl)furan-2-yl)methylene)thiazolidin-4-one | 301687-87-6 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY119648A10MG
Encompass_Preferred
(E/Z)-CCR-11 is a small-molecule research compound that inhibits CD38 cyclase activity and has been reported to increase cellular NAD+ levels and interferon-γ in cellular assays.
  • Selective CD38 inhibitor with IC50 20.8 μM.
  • Promotes increases in cellular NAD+ levels.
  • Associated with increased interferon-γ in cellular studies.
  • Molecular formula C15H8F3NO2S2, molecular weight 355.35.
  • High purity suitable for research use (98.0%).
  • Available in small research pack sizes (for example, 5 mg and 10 mg vials; larger quantities available by quote).
Catalog No. 50-002-33625